4-Hydroxypyridine, 95%
4-Hydroxypyridine, 95%
4-Hydroxypyridine, 95%
Thermo Scientific Chemicals

4-Hydroxypyridine, 95%

CAS: 626-64-2 | C5H5NO | 95.10 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 348831000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS626-64-2
IUPAC Name1,4-dihydropyridin-4-one
Molecular FormulaC5H5NO
InChI KeyGCNTZFIIOFTKIY-UHFFFAOYSA-N
SMILESO=C1C=CNC=C1
さらに表示
Titration with HClO4>=94 %
HPLC>=94 %
Melting point139°C to 151°C
Appearance (Color)White to yellow to beige to light brown
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only