Calcium chloride, 93+%, extra pure, powder, anhydrous
Calcium chloride, 93+%, extra pure, powder, anhydrous
Calcium chloride, 93+%, extra pure, powder, anhydrous
Thermo Scientific Chemicals

Calcium chloride, 93+%, extra pure, powder, anhydrous

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol
数量:
25 g
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 349610100
価格(JPY)
-
数量:
10 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10043-52-4
IUPAC Namecalcium dichloride
Molecular FormulaCaCl2
InChI KeyUXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ca++]
さらに表示
Appearance (Color)White
Iron (Fe)=<50 ppm
Barium (Ba)=<200 ppm
Sulfate (SO4)=<500 ppm
Magnesium (Mg)=<1000 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only