GlutaMAX™ Supplement, 20 x 100 mL - FAQs
View additional product information for GlutaMAX™ Supplement - FAQs (35050087)
3 product FAQs found
Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?
Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?
In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between Gibco GlutaMAX and glutamine supplements?
GlutaMAX Supplement is an improved cell culture supplement that can be used as a direct substitute for L-glutamine in your cell culture medium. Please go here to read more about the properties of GlutaMAX supplement (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/media-supplements/glutamax-media/glutamax-vs-glutamine.html).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.