4-chlorophenylacetone, 98%
4-chlorophenylacetone, 98%
4-chlorophenylacetone, 98%
Thermo Scientific Chemicals

4-chlorophenylacetone, 98%

CAS: 5586-88-9 | C9H9ClO | 168.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 350700250
Price (USD)
250.00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
250.00
Each
Chemical Identifiers
CAS5586-88-9
IUPAC Name1-(4-chlorophenyl)propan-2-one
Molecular FormulaC9H9ClO
InChI KeyWEJRYKSUUFKMBC-UHFFFAOYSA-N
SMILESCC(=O)CC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5325 to 1.5345 (20°C, 589 nm)
Appearance (Color)Clear light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only