2-Chloro-3-cyanopyridine, 98%
2-Chloro-3-cyanopyridine, 98%
2-Chloro-3-cyanopyridine, 98%
Thermo Scientific Chemicals

2-Chloro-3-cyanopyridine, 98%

CAS: 6602-54-6 | C6H3ClN2 | 138.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 352080250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6602-54-6
IUPAC Name2-chloropyridine-3-carbonitrile
Molecular FormulaC6H3ClN2
InChI KeyJAUPUQRPBNDMDT-UHFFFAOYSA-N
SMILESClC1=NC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Melting point105°C to 110°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only