1-Butyl-3-methylimidazolium chloride, 98+%
1-Butyl-3-methylimidazolium chloride, 98+%
1-Butyl-3-methylimidazolium chloride, 98+%
1-Butyl-3-methylimidazolium chloride, 98+%
Thermo Scientific Chemicals

1-Butyl-3-methylimidazolium chloride, 98+%

CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 354095000
Price (EUR)
585,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
585,00
Each
Chemical Identifiers
CAS79917-90-1
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium chloride
Molecular FormulaC8H15ClN2
InChI KeyFHDQNOXQSTVAIC-UHFFFAOYSA-M
SMILES[Cl-].CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.0 %
Water=<1.0 % (K.F.)
Appearance (Color)Cream to yellow to orange
Infrared spectrumConforms
Appearance (Form)Powder or crystals or chunks or fused solid

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Ionic liquid
RUO – Research Use Only