Methyl 5-bromo-2-furoate, 98%
Methyl 5-bromo-2-furoate, 98%
Methyl 5-bromo-2-furoate, 98%
Thermo Scientific Chemicals

Methyl 5-bromo-2-furoate, 98%

CAS: 2527-99-3 | C6H5BrO3 | 205.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 355750250
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2527-99-3
IUPAC Namemethyl 5-bromofuran-2-carboxylate
Molecular FormulaC6H5BrO3
InChI KeyFBPIDMAELBIRLE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Br)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Melting point63°C to 68°C
Appearance (Form)Crystalline powder
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only