Thermo Scientific Chemicals

3-Trifluoromethylphenylboronic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 359750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Trifluoromethylphenylboronic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 359750050
5 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
1423-26-3
IUPAC Name
[3-(trifluoromethyl)phenyl]boronic acid
Molecular Formula
C7H6BF3O2
InChI Key
WOAORAPRPVIATR-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC(=C1)C(F)(F)F
Infrared spectrum
Conforms
Melting point
161°C to 167°C
HPLC
>=97.0 %
Appearance (Color)
White to light yellow
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text