Thermo Scientific Chemicals

2-Methyl-2-pentenal, 97%, Thermo Scientific Chemicals

Catalog number: 359961000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methyl-2-pentenal, 97%, Thermo Scientific Chemicals

Catalog number: 359961000
100 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
623-36-9
IUPAC Name
(2E)-2-methylpent-2-enal
Molecular Formula
C6H10O
InChI Key
IDEYZABHVQLHAF-GQCTYLIASA-N
SMILES
CCC=C(/C)C=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=96.0 %
Refractive index
1.4470 to 1.4550 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text