(S)-4-Fluorophenylglycine, 95%, (98% ee), Thermo Scientific Chemicals
(S)-4-Fluorophenylglycine, 95%, (98% ee), Thermo Scientific Chemicals
(S)-4-Fluorophenylglycine, 95%, (98% ee), Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-4-Fluorophenylglycine, 95%, (98% ee), Thermo Scientific Chemicals

CAS: 19883-57-9 | Molecular Formula: C8H8FNO2 | Molecular Weight: 169.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 362050010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19883-57-9
IUPAC Name(2S)-2-azaniumyl-2-(4-fluorophenyl)acetate
Molecular FormulaC8H8FNO2
InChI KeyJKFYKCYQEWQPTM-ZETCQYMHSA-N
SMILES[NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
HPLC>=94 %
Enantiomeric excess>=97.5 % (HPLC)
Appearance (Form)Powder
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only