(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%
(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%
(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%
Thermo Scientific Chemicals

(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, 95%

CAS: 79200-57-0 | C6H13NO3·HCl | 183.63 g/mol
数量:
250 mg
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 362520050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79200-57-0
IUPAC Name(1R,2S,3R,4R)-2,3-dihydroxy-4-(hydroxymethyl)cyclopentan-1-aminium
Molecular FormulaC6H14NO3
InChI KeyFHNKBDPGQXLKRW-KAZBKCHUSA-O
SMILES[NH3+][C@@H]1C[C@H](CO)[C@@H](O)[C@H]1O
さらに表示
Appearance (Color)White to off-white
Infrared spectrumConforms
Specific optical rotation-16° to -20° (20°C, 589 nm) (c=1, methanol)
Appearance (Form)Crystalline powder and/or chunks
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only