Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack
Thermo Scientific™

Nalgene™ PPCO Amber Micro Packaging Vials: Nonsterile, Bulk Pack

Protect light-sensitive reagents with these PPCO amber micro packaging vials featuring conical bottoms that allow recovery of entire contents.
製品番号(カタログ番号)容量(メートル法)
362805-00202 mL
362805-00050.5 mL
製品番号(カタログ番号) 362805-0020
価格(JPY)
42,500
キャンペーン価格
Ends: 25-Dec-2026
53,140
割引額 10,640 (20%)
Case of 1000
容量(メートル法):
2 mL
Protect light sensitive reagents with Thermo Scientific™ Nalgene™ PPCO Amber Micro Packaging Vials. These noncytotoxic and nonpyrogenic vials are molded from high purity, low-metal content polypropylene copolymer resin. They are skirted and feature a conical bottom, allowing recovery of entire contents.
Features:
  • Vials and closures (listed separately) pressure-tested together at 7.5 psig
  • USP Class VI compliant; nonpyrogenic
  • Can be centrifuged at 13,000 x g
  • Colored closures packaged separately
  • Nonsterile
  • Leakproof† when used with Nalgene closures

Ordering Information:

Vials are bulk packed without closures, 500 per inner bag, 1,000 per case. Order nonsterile closures separately (Cat. Nos. 362821, 362825).

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

仕様
認証/コンプライアンスISO 13485:2016
クロージャー材質PPCO
クロージャーサイズ11 mm
概要Micro Packaging Vials
直径(メートル法)内径8 mm
高さ(メートル法)49 mm
箱あたりの入数1000
パックあたり入数500
製品タイプVial
容量(メートル法)合計2 mL
キャップタイプScrew Cap (Not Included)
色Amber
直径(メートル法)外径13 mm
材料PPCO
数量500 Bag, 1000 Cs.
無菌性Non-sterile
容量(メートル法)2 mL
Unit SizeCase of 1000

よくあるご質問(FAQ)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.