1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 872-50-4 | C5H9NO | 99.133 g/mol
数量:
1 L
2.5 L
パッケージ:
AcroSeal™ Glass Bottle
製品番号(カタログ番号) 364380010
価格(JPY)
-
数量:
1 L
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
さらに表示
GC>=99.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
Color scale=<50 APHA

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only