Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 364411000
價格 (TWD)
-
Quantity:
100 mL
Request bulk or custom format
化學識別
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
Appearance (Color)Clear colorless
GC>=99.8 %
Acidity (HCl)=<0.002 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only