2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 364670010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
Appearance (Form)Liquid
GC>=99.4 %
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only