Boron tribromide, 99+%, Thermo Scientific Chemicals
Boron tribromide, 99+%, Thermo Scientific Chemicals
Boron tribromide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron tribromide, 99+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
36521025025gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 365210250
Price (EUR)
49,65
Online exclusive
55,40
Save 5,75 (10%)
Each
Quantity:
25g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
49,65
Online exclusive
55,40
Save 5,75 (10%)
Each
Chemical Identifiers
CAS10294-33-4
IUPAC Nametribromoborane
Molecular FormulaBBr3
InChI KeyILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILESBrB(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Heavy metals (as Pb)=<100 ppm
Impurity=<0.2 % Free bromine (Br2)
Sulfur (S)=<100 ppm
Titration99.0 to 101.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acid
RUO – Research Use Only