2-Quinolinecarboxaldehyde, 97%
2-Quinolinecarboxaldehyde, 97%
2-Quinolinecarboxaldehyde, 97%
2-Quinolinecarboxaldehyde, 97%
Thermo Scientific Chemicals

2-Quinolinecarboxaldehyde, 97%

CAS: 5470-96-2 | C10H7NO | 157.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 370170250
Price (EUR)
244,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
244,00
Each
Add to cart
Chemical Identifiers
CAS5470-96-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Appearance (Form)Crystalline powder or crystals
Melting point66°C to 71°C
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only