Tributylphenyltin
Tributylphenyltin
Tributylphenyltin
Thermo Scientific Chemicals

Tributylphenyltin

CAS: 960-16-7 | C18H32Sn | 367.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 370230010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS960-16-7
IUPAC Name(2,3,4-tributylphenyl)stannanylidyne
Molecular FormulaC18H29Sn
InChI KeyZMUNITMSSGNSOT-UHFFFAOYSA-N
SMILESCCCCC1=CC=C([Sn])C(CCCC)=C1CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.5150 to 1.5170 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only