Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution
Thermo Scientific Chemicals

Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution

CAS: 75-50-3 | C3H9N | 59.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 370780025
Price (EUR)
178,00
Each
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
178,00
Each
Add to cart
Chemical Identifiers
CAS75-50-3
IUPAC Nametrimethylamine
Molecular FormulaC3H9N
InChI KeyGETQZCLCWQTVFV-UHFFFAOYSA-N
SMILESCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl48 to 52 %
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only