2-Ethylhexyl methacrylate, 99%, stabilized
2-Ethylhexyl methacrylate, 99%, stabilized
2-Ethylhexyl methacrylate, 99%, stabilized
Thermo Scientific Chemicals

2-Ethylhexyl methacrylate, 99%, stabilized

CAS: 688-84-6 | C12H22O2 | 198.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 371170010
Price (USD)
89.65
Special offer
Online exclusive
Ends: 28-Dec-2026
105.00
Save 15.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
89.65
Special offer
Online exclusive
Ends: 28-Dec-2026
105.00
Save 15.35
Each
Chemical Identifiers
CAS688-84-6
IUPAC Name2-ethylhexyl 2-methylprop-2-enoate
Molecular FormulaC12H22O2
InChI KeyWDQMWEYDKDCEHT-UHFFFAOYNA-N
SMILESCCCCC(CC)COC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=98.5 %
Color scale=<15 APHA
Infrared spectrumConforms
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only