Quinaldine red, pure
Quinaldine red, pure
Quinaldine red, pure
Thermo Scientific Chemicals

Quinaldine red, pure

CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 377510250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS117-92-0
IUPAC Name2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
Molecular FormulaC21H23IN2
InChI KeyJOLANDVPGMEGLK-UHFFFAOYSA-M
SMILES[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
さらに表示
Appearance (Color)Grey to green-black to dark blue
Infrared spectrumConforms
Appearance (Form)Powder
UV spectrumComplies
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only