Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%
Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%
Thermo Scientific Chemicals

Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate, 98%

CAS: 80049-61-2 | C11H14F6N3PRu | 434.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 377890010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS80049-61-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Appearance (Form)Powder
Infrared spectrumConforms
Assay>=97.5 % (ex Ru)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only