Bis(triethylphosphine)palladium(II) chloride, Thermo Scientific Chemicals
Bis(triethylphosphine)palladium(II) chloride, Thermo Scientific Chemicals
Bis(triethylphosphine)palladium(II) chloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(triethylphosphine)palladium(II) chloride, Thermo Scientific Chemicals

CAS: 28425-04-9 | Molecular Formula: C12H30Cl2P2Pd | Molecular Weight: 413.64 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3779700101 gGlass bottle
Catalog number 377970010
Price (USD)
161.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
161.00
Each
Chemical Identifiers
CAS28425-04-9
IUPAC Namedichloropalladium; bis(triethylphosphane)
Molecular FormulaC12H30Cl2P2Pd
InChI KeyULYNIEUXPCUIEL-UHFFFAOYSA-L
SMILESCl[Pd]Cl.CCP(CC)CC.CCP(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Palladium (Pd)>=24.9 %
Appearance (Color)Yellow

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only