n-Butyllithium, 2.7M solution in toluene, AcroSeal™
n-Butyllithium, 2.7M solution in toluene, AcroSeal™
n-Butyllithium, 2.7M solution in toluene, AcroSeal™
Thermo Scientific Chemicals

n-Butyllithium, 2.7M solution in toluene, AcroSeal™

CAS: 109-72-8 | C4H9Li | 64.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 378938000
Price (TWD)
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS109-72-8
IUPAC Namebutyllithium
Molecular FormulaC4H9Li
InChI KeyMZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES[Li]CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Active base2.62 to 2.75 M
Appearance (Color)Yellow to orange
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Free base=<1.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only