Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Lithium hydroxide, 98%, pure, anhydrous
Thermo Scientific Chemicals

Lithium hydroxide, 98%, pure, anhydrous

CAS: 1310-65-2 | HLiO | 23.95 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 379180025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1310-65-2
IUPAC Namelithium(1+) hydroxide
Molecular FormulaHLiO
InChI KeyWMFOQBRAJBCJND-UHFFFAOYSA-M
SMILES[Li+].[OH-]
さらに表示
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Titration with HCl>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only