5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%
5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%
5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%
Thermo Scientific Chemicals

5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%

CAS: 22934-51-6 | C11H12O3 | 192.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 379360100
Price (USD)
92.30
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
92.30
Each
Chemical Identifiers
CAS22934-51-6
IUPAC Name2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzaldehyde
Molecular FormulaC11H12O3
InChI KeyFDHXEIOBIOVBEN-UHFFFAOYSA-N
SMILESCOC1=CC(CC=C)=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Light yellow
Melting point48°C to 52°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only