Cyclohexylacetylene, 98%
Cyclohexylacetylene, 98%
Cyclohexylacetylene, 98%
Thermo Scientific Chemicals

Cyclohexylacetylene, 98%

CAS: 931-48-6 | C8H12 | 108.18 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 380790050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS931-48-6
IUPAC Nameethynylcyclohexane
Molecular FormulaC8H12
InChI KeySSDZYLQUYMOSAK-UHFFFAOYSA-N
SMILESC#CC1CCCCC1
さらに表示
Refractive index1.4530 to 1.4550 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only