Methyl 5-iodosalicylate 99%, Thermo Scientific Chemicals
Methyl 5-iodosalicylate 99%, Thermo Scientific Chemicals
Methyl 5-iodosalicylate 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 5-iodosalicylate 99%, Thermo Scientific Chemicals

CAS: 4068-75-1 | C8H7IO3 | 278.045 g/mol
製品番号(カタログ番号)数量パッケージ
38177050050 gGlass bottle
製品番号(カタログ番号) 381770500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4068-75-1
IUPAC Namemethyl 2-hydroxy-5-iodobenzoate
Molecular FormulaC8H7IO3
InChI KeyNRSWJTRJHPRZMH-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(I)=CC=C1O
さらに表示
Appearance (Color)White
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder
Melting point73°C to 78°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only