1,3-Dichloroisoquinoline, 97%
1,3-Dichloroisoquinoline, 97%
1,3-Dichloroisoquinoline, 97%
Thermo Scientific Chemicals

1,3-Dichloroisoquinoline, 97%

CAS: 7742-73-6 | C9H5Cl2N | 198.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 382410050
Price (USD)
234.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
234.00
Each
Chemical Identifiers
CAS7742-73-6
IUPAC Name1,3-dichloroisoquinoline
Molecular FormulaC9H5Cl2N
InChI KeyBRGZEQXWZWBPJH-UHFFFAOYSA-N
SMILESClC1=NC(Cl)=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)White to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point120°C to 125°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only