Nalgene™ Certified Clean PETG Biotainer™ Media Bottle
Thermo Scientific™
Nalgene™ Certified Clean PETG Biotainer™ Media Bottle
For Use With: Specifically designed for pharmaceutical and biological manufacturing.; Color: Clear
製品番号(カタログ番号)容量(メートル法)
383110-351 L
383005-70500 mL
製品番号(カタログ番号) 383110-35
価格(JPY)
201,100
Each
容量(メートル法):
1 L
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Media Bottles reduce the risk of carry-over contamination.
For Research Use Only. Not for use in diagnostic procedures.
仕様
オートクレーブ可能Not Autoclavable
使用対象(アプリケーション)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
微粒子USP <788> (Low particulate)
箱あたりの入数35/Cs.
ワークフローステップUpstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
容量(メートル法)1 L
Clear
材料PET
製品タイプSquare Bottle
無菌性Sterile
Unit SizeEach