Samarium(III) trifluoromethanesulfonate, 98%
Samarium(III) trifluoromethanesulfonate, 98%
Samarium(III) trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Samarium(III) trifluoromethanesulfonate, 98%

CAS: 52093-28-4 | C3F9O9S3Sm | 597.55 g/mol
数量:
1 g
10 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 383170100
価格(JPY)
-
数量:
10 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS52093-28-4
IUPAC Namesamarium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9O9S3Sm
InChI KeyDDCWGUIPLGMBPO-UHFFFAOYSA-K
SMILES[Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
さらに表示
Infrared spectrumConforms
Appearance (Color)White
Elemental Analysis24.65 to 25.66 % (Sm)
Appearance (Form)Powder
Elemental Analysis5.91 to 6.15 % (C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only