4-Bromo-N-methylaniline, 97%
4-Bromo-N-methylaniline, 97%
4-Bromo-N-methylaniline, 97%
Thermo Scientific Chemicals

4-Bromo-N-methylaniline, 97%

CAS: 6911-87-1 | C7H8BrN | 186.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 383200050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6911-87-1
IUPAC Name4-bromo-N-methylaniline
Molecular FormulaC7H8BrN
InChI KeyAYVPVDWQZAAZCM-UHFFFAOYSA-N
SMILESCNC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.6110 to 1.6150 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to brown to dark purple
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only