Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous
Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous
Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous
Thermo Scientific Chemicals

Gadolinium(III) chloride, 99.9%, (trace metal basis), anhydrous

CAS: 10138-52-0 | Cl3Gd | 263.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 383560050
Price (USD)
192.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
192.00
Each
Chemical Identifiers
CAS10138-52-0
IUPAC Namegadolinium(3+) trichloride
Molecular FormulaCl3Gd
InChI KeyMEANOSLIBWSCIT-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Total trace metal impurities=<1000 ppm
Titration Complexometric58.2 to 61.1 % (Gd)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only