n-Hexane, for analysis, contains appr. 95% n-Hexane
n-Hexane, for analysis, contains appr. 95% n-Hexane
n-Hexane, for analysis, contains appr. 95% n-Hexane
Thermo Scientific Chemicals

n-Hexane, for analysis, contains appr. 95% n-Hexane

CAS: 110-54-3 | C6H14 | 86.18 g/mol
数量:
1 L
2.5 L
25 L
製品番号(カタログ番号) 383800250
価格(JPY)
-
数量:
25 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-54-3
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
さらに表示
Hydrocarbons=<0.1 % Aromatic hydrocarbons
Color scale=<10 APHA
Water=<0.02 % (K.F.)
Bromine number=<0.5
Copper (Cu)=<0.02 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only