Isohexane, for HPLC, contains <5% n-Hexane
Isohexane, for HPLC, contains <5% n-Hexane
Isohexane, for HPLC, contains <5% n-Hexane
Thermo Scientific Chemicals

Isohexane, for HPLC, contains <5% n-Hexane

Green features
CAS: 73513-42-5 | C6H14 | 86.18 g/mol
製品番号(カタログ番号) 383810010
価格(JPY)
-
数量:
1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS73513-42-5
IUPAC Name2-methylpentane
Molecular FormulaC6H14
InChI KeyAFABGHUZZDYHJO-UHFFFAOYSA-N
SMILESCCCC(C)C
さらに表示
UVat 220 nm A: =<0.07
UVat 240 nm A: =<0.02
UVat 260 nm A: =<0.005
Filtered to 0.2 micrometerFiltered
Appearance (Color)Clear colorless
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only