4-Aminomethyltetrahydropyran, 97%
4-Aminomethyltetrahydropyran, 97%
4-Aminomethyltetrahydropyran, 97%
Thermo Scientific Chemicals

4-Aminomethyltetrahydropyran, 97%

CAS: 130290-79-8 | C6H13NO | 115.17 g/mol
製品番号(カタログ番号) 385210050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS130290-79-8
IUPAC Name(oxan-4-yl)methanaminium
Molecular FormulaC6H14NO
InChI KeyIPBPLHNLRKRLPJ-UHFFFAOYSA-O
SMILES[NH3+]CC1CCOCC1
さらに表示
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4640 to 1.4670 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only