Pyridine, 99+%, for spectroscopy
Pyridine, 99+%, for spectroscopy
Pyridine, 99+%, for spectroscopy
Thermo Scientific Chemicals

Pyridine, 99+%, for spectroscopy

CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass Bottle
Catalog number 386560250
Price (TWD)
-
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Residue after evaporation=<0.001 %
Color scale=<10 APHA
Water=<0.05 %
UVat 335 nm A: =<0.301
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only