4-Penten-2-ol, 96%
4-Penten-2-ol, 96%
4-Penten-2-ol, 96%
Thermo Scientific Chemicals

4-Penten-2-ol, 96%

CAS: 625-31-0 | C5H10O | 86.13 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 387430050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS625-31-0
IUPAC Namepent-4-en-2-ol
Molecular FormulaC5H10O
InChI KeyZHZCYWWNFQUZOR-UHFFFAOYNA-N
SMILESCC(O)CC=C
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=95 %
Refractive index1.4230 to 1.4250 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only