Hydrochloric acid, for analysis, ca. 32% solution in water
Hydrochloric acid, for analysis, ca. 32% solution in water
Hydrochloric acid, for analysis, ca. 32% solution in water
Thermo Scientific Chemicals

Hydrochloric acid, for analysis, ca. 32% solution in water

CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3893000252.5 LGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 389300025
Price (EUR)
58,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
58,00
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ign. (sulfated)=<0.0005 %
Calcium (Ca)=<0.5 ppm
Chromium (Cr)=<0.02 ppm
Magnesium (Mg)=<0.1 ppm
Sodium (Na)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only