Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water
Thermo Scientific Chemicals

Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water

CAS: 124-64-1 | C4H12ClO4P | 190.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
Glass bottle
Catalog number 389441000
Price (USD)
66.90
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
66.90
Each
Chemical Identifiers
CAS124-64-1
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Solution
Appearance (Color)Colorless to yellow-green
Titration Argentometric75 to 85 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only