Dimethylaminoacetaldehyde dimethylacetal, 97%
Dimethylaminoacetaldehyde dimethylacetal, 97%
Dimethylaminoacetaldehyde dimethylacetal, 97%
Thermo Scientific Chemicals

Dimethylaminoacetaldehyde dimethylacetal, 97%

CAS: 38711-20-5 | C6H15NO2 | 133.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 390910010
Price (EUR)
27,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
27,00
Each
Chemical Identifiers
CAS38711-20-5
IUPAC Name(2,2-dimethoxyethyl)dimethylamine
Molecular FormulaC6H15NO2
InChI KeyHUYAEQCJNXODLQ-UHFFFAOYSA-N
SMILESCOC(CN(C)C)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4080 to 1.4120 (20°C, 589 nm)
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only