Chloro(triphenylphosphine)gold(I), 99%
Chloro(triphenylphosphine)gold(I), 99%
Chloro(triphenylphosphine)gold(I), 99%
Thermo Scientific Chemicals

Chloro(triphenylphosphine)gold(I), 99%

CAS: 14243-64-2 | C18H15AuClP | 494.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 391610010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14243-64-2
IUPAC Namechlorogold; triphenylphosphane
Molecular FormulaC18H15AuClP
InChI KeyIFPWCRBNZXUWGC-UHFFFAOYSA-M
SMILESCl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Assay>=98.5 % (ex Au)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only