Indole-5-carboxaldehyde, 98+%
Indole-5-carboxaldehyde, 98+%
Indole-5-carboxaldehyde, 98+%
Thermo Scientific Chemicals

Indole-5-carboxaldehyde, 98+%

CAS: 1196-69-6 | C9H7NO | 145.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 391630010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1196-69-6
IUPAC Name1H-indole-5-carbaldehyde
Molecular FormulaC9H7NO
InChI KeyADZUEEUKBYCSEY-UHFFFAOYSA-N
SMILESO=CC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point98°C to 103°C
Appearance (Color)Orange to tan
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only