Thermo Scientific Chemicals

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II), Thermo Scientific Chemicals

Catalog number: 392140050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II), Thermo Scientific Chemicals

Catalog number: 392140050
5 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
16971-33-8
IUPAC Name
chloro(hydrido)rutheniumiumylium; methanidylidyneoxidanium; tris(triphenylphosphane)
Molecular Formula
C55H46ClOP3Ru
InChI Key
IMPHYUNMDWWJAH-UHFFFAOYSA-M
SMILES
Cl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Powder or crystals
Appearance (Color)
Cream to tan to brown
Ruthenium (Ru)
>=10.3 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text