(±)-1-Phenyl-2-propyn-1-ol, 98+%
(±)-1-Phenyl-2-propyn-1-ol, 98+%
(±)-1-Phenyl-2-propyn-1-ol, 98+%
(±)-1-Phenyl-2-propyn-1-ol, 98+%
Thermo Scientific Chemicals

(±)-1-Phenyl-2-propyn-1-ol, 98+%

CAS: 4187-87-5 | C9H8O | 132.16 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 392760250
価格(JPY)
-
見積もりを依頼する
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4187-87-5
IUPAC Name1-phenylprop-2-yn-1-ol
Molecular FormulaC9H8O
InChI KeyUIGLAZDLBZDVBL-UHFFFAOYNA-N
SMILESOC(C#C)C1=CC=CC=C1
さらに表示
Appearance (Color)Clear colorless to yellow
GC>=98.0 %
Appearance (Form)Liquid
Refractive index1.5450 to 1.5530 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only