2-(2-Bromophenyl)-1,3-dioxolane, 98%
2-(2-Bromophenyl)-1,3-dioxolane, 98%
2-(2-Bromophenyl)-1,3-dioxolane, 98%
Thermo Scientific Chemicals

2-(2-Bromophenyl)-1,3-dioxolane, 98%

CAS: 34824-58-3 | C9H9BrO2 | 229.07 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 393240250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS34824-58-3
IUPAC Name2-(2-bromophenyl)-1,3-dioxolane
Molecular FormulaC9H9BrO2
InChI KeyIWSGKSUCFVOWQU-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1C1OCCO1
さらに表示
Refractive index1.5670 to 1.5700 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only