Product Image
Thermo Scientific Chemicals

1,3-Dimethylbarbituric acid, 98%

CAS: 769-42-6 | C6H8N2O3 | 156.14 g/mol
製品番号(カタログ番号) 393281000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS769-42-6
IUPAC Name1,3-dimethyl-1,3-diazinane-2,4,6-trione
Molecular FormulaC6H8N2O3
InChI KeyVVSASNKOFCZVES-UHFFFAOYSA-N
SMILESCN1C(=O)CC(=O)N(C)C1=O
さらに表示
Appearance (Color)White to yellow to light brown
Melting point121°C to 125°C
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only