Raney™-Nickel, Activated Catalyst, 50% slurry in water
Raney™-Nickel, Activated Catalyst, 50% slurry in water
Raney™-Nickel, Activated Catalyst, 50% slurry in water
Thermo Scientific Chemicals

Raney™-Nickel, Activated Catalyst, 50% slurry in water

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
395925000500 g
Catalog number 395925000
Price (USD)
204.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
204.00
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Aluminium (Al)=<8 %
pH8.5 to 12.0 (of supernatant)
Appearance (Form)Slurry in water
Nickel (Ni)>=92 %
Appearance (Color)Dark grey to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only