Methylamine, 33 wt.% solution in absolute ethanol, AcroSeal™
Thermo Scientific Chemicals

Methylamine, 33 wt.% solution in absolute ethanol, AcroSeal™

CAS: 74-89-5 | CH5N | 31.06 g/mol
数量:
100 mL
800 mL
製品番号(カタログ番号) 396731000
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS74-89-5
IUPAC Namemethanamine
Molecular FormulaCH5N
InChI KeyBAVYZALUXZFZLV-UHFFFAOYSA-N
SMILESCN
さらに表示
Titration with HClO431.0 to 35.0 %
Appearance (Color)Clear colorless
Appearance (Form)Solution
Infrared spectrumConforms
Water=<1.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use