2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%
2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%
2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%

CAS: 22195-47-7 | C6H13NO2 | 131.18 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 397410050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS22195-47-7
IUPAC Name1-(2,2-dimethyl-1,3-dioxolan-4-yl)methanamine
Molecular FormulaC6H13NO2
InChI KeyHXOYWCSTHVTLOW-UHFFFAOYNA-N
SMILESCC1(C)OCC(CN)O1
さらに表示
Infrared spectrumConforms
Refractive index1.4370 to 1.4390 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
Appearance (Color)Colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only